Orders LinkContactUsLinkComponent
TWD Barcoded Autosampler Vials
TWD Barcoded Autosampler Vials
Vial and cap kit, 2.00 ml, 12×32 mm, Glass, Amber, Yes, Red rubber/PTFE
Catalog # 75780-316
Supplier:   TWD Scientific
Supplier #:  GV1232A-P-2DK
TWD Barcoded Autosampler Vials
Catalog # 75780-316
Supplier:  TWD Scientific
TWD Barcoded Autosampler Vials
TWD Barcoded Autosampler Vials
Vial and cap kit, 2.00 ml, 12×32 mm, Glass, Amber, Yes, Red rubber/PTFE
Price
196,06 $
Log in for availability
Case of 100
Product Details & Documents
TWD Barcoded Autosampler Vials with 2D barcode.
  • Vials feature writing patch and graduations
  • Glass construction
  • Screw-top 9-425 GPI thread finish
TWD barcoded autosampler kits are a cost-effective option if you're looking for a easy, hassle-free option to track your samples your samples.
Delivery information: Convenient Kit includes black PP cap and red rubber/PTFE Septa; available as vial and cap kit or cap only.
Specifications
  • Cap included:
    Yes
  • Septum:
    Red rubber/PTFE
  • Capacity:
    2.00 ml
  • Color:
    Amber
  • Description:
    Vial and cap kit
  • Material:
    Glass
  • Diameter ×H:
    12×32 mm