Order Entry
Canada
Orders LinkContactUsLinkComponent
Rigid-Lock Quickberm, Justrite®

Rigid-Lock Quickberm, Justrite®

Quickberm Rigid-Lock, 144 W x 12 H x 240 D", 152 W x 12 H x 248 L"
  10861-148
 :   Justrite
Supplier #:  28526
Rigid-Lock Quickberm, Justrite®
  10861-148
 :  Justrite
Rigid-Lock Quickberm, Justrite®
Rigid-Lock Quickberm, Justrite®

Quickberm Rigid-Lock, 144 W x 12 H x 240 D", 152 W x 12 H x 248 L"

 
$5,425.56
   
Each (1,795US Gal)
 

Ergonomic design provides 100% inside berm accessibility with an unobstructed side-to-side floor space for vehicle tires, foot traffic, and cleaning.

  • Single-piece construction - patented Rigid-Lock wall-support system integral to berm provides unobstructed work space
  • Quick and easy setup - no assembly required
  • Durable EnGuard™V, modified PVC coated fabric withstands long-term UV exposure

Exceptionally strong structural integrity includes a reinforced CriticalCorner™ design featuring wraparound diagonal radio frequency welds to prevent billowing, unwrapping, or sagging of the corners during full containment for fluid-tight reliability

The built-in Rigid-Lock wall supports maximize usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter. The device only folds down one way, inward towards the inside of the berm. The one-piece design folds down flat and remains integral to the sidewall. Supports resist extreme temperatures (-50 to 160° F). When in the down position, they can withstand vehicle weights in excess of 11,000 lbs (5,000 kg) at each tire.

Sidewalls lower quickly for easy drive-in and out. Portable design folds down quickly for storage and transport.

 
  • Description:
    Quickberm Rigid-Lock
  • Interior dimensions:
    144 W x 12 H x 240 D"
  • Ext. dimensions:
    152 W x 12 H x 248 L"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.