Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 $
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SWAB OPTI-TRANZ POLYSTYTRENE HANDLE BG50
Catalog # 10860-996
Supplier:
Puritan Medical Products
Expand image
SWAB OPTI-TRANZ POLYSTYTRENE HANDLE BG50
Catalog # 10860-996
Supplier:
Puritan Medical Products
Supplier Number:
LA-62PR
Related Information
Documentation
Product Literature
Puritan Medical Products Opti-Tranz Plus Brochure
Documentation
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning