Orders LinkContactUsLinkComponent
Inserts for Versa Vials™, Restek
Inserts for Versa Vials™, Restek
Versa vial insert, 0.25 ml, Flat bottom, Glass, No
Catalog # 10854-368
Supplier:   Restek
Supplier #:  22711
Inserts for Versa Vials™, Restek
Catalog # 10854-368
Supplier:  Restek
Inserts for Versa Vials™, Restek
Inserts for Versa Vials™, Restek
Versa vial insert, 0.25 ml, Flat bottom, Glass, No
Price
$40.68
New Customer Promotional Price
Log in for availability
Pack of 100
Product Details & Documents
Glass and polypropylene Versa Vial™ inserts are compatible with Versa Vials™ of various sizes.
  • 250 µl volume vial compatibility
  • Glass or polypropylene construction
Inserts for Versa Vials™ are made of rugged polypropylene or glass and feature a flat-bottomed design or flanges.
Specifications
  • Cap included:
    No
  • Capacity:
    0.25 ml
  • Description:
    Versa vial insert
  • Material:
    Glass
  • Bottom style:
    Flat bottom