Orders LinkContactUsLinkComponent
QuickSplit Post-Column Flow Splitters, Restek
Replacement Fixed Flow Splitter Resistor Set, Split Ratio 20:1
  10853-118
 :   Restek
 :  25333
QuickSplit Post-Column Flow Splitters, Restek
  10853-118
 :  Restek
QuickSplit Post-Column Flow Splitters, Restek
QuickSplit Post-Column Flow Splitters, Restek
Replacement Fixed Flow Splitter Resistor Set, Split Ratio 20:1
$2,079.10
Each
Fluid resistor technology eliminates adjustments to capillary tubing for optimizing split ratio. Wide range of interchangeable resistors available.
  • Split ratio not affected by changes in viscosity or pressure
  • Low dead volume - negligible effect on analyte bandwidth
  • High operating pressure limit: Fixed flow splitters 10000 psi (68,948 kPa) and adjustable flow splitter 5000 psi (34,474 kPa)
  • Total flow: 0.1 to 5.0 ml/min
Adjustable Flow Splitter: Adjustable metering valve gives convenient control of split ratio.
 : *QuickSplit post-column flow splitter serial number required for replacement resistor set to ensure compatibility. Every resistor is manufactured per serial number. The serial number is located on top of the splitter.
  • Description:
    Replacement Fixed Flow Splitter Resistor Set, Split Ratio 20:1