Order Entry
Canada
Orders LinkContactUsLinkComponent
QuickSplit Post-Column Flow Splitters, Restek

QuickSplit Post-Column Flow Splitters, Restek

QuickSplit Binary Fixed Flow Splitter, Split Ratio 100:1 Post Column
Catalog # 10853-110
Supplier:   Restek
Supplier #:  25326
QuickSplit Post-Column Flow Splitters, Restek
Catalog # 10853-110
Supplier:  Restek
QuickSplit Post-Column Flow Splitters, Restek
QuickSplit Post-Column Flow Splitters, Restek

QuickSplit Binary Fixed Flow Splitter, Split Ratio 100:1 Post Column

Price
$4,054.95
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Fluid resistor technology eliminates adjustments to capillary tubing for optimizing split ratio. Wide range of interchangeable resistors available.

  • Split ratio not affected by changes in viscosity or pressure
  • Low dead volume - negligible effect on analyte bandwidth
  • High operating pressure limit: Fixed flow splitters 10000 psi (68,948 kPa) and adjustable flow splitter 5000 psi (34,474 kPa)
  • Total flow: 0.1 to 5.0 ml/min

Adjustable Flow Splitter: Adjustable metering valve gives convenient control of split ratio.

Ordering information: *QuickSplit post-column flow splitter serial number required for replacement resistor set to ensure compatibility. Every resistor is manufactured per serial number. The serial number is located on top of the splitter.

Specifications
  • Description:
    QuickSplit Binary Fixed Flow Splitter, Split Ratio 100:1 Post Column
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.