VWR® Dulbecco's Phosphate Buffered Saline (DPBS)
DPBS (1X) w/out Ca, w/out Mg, 1000 ml
Supplier #: 02-0119-1000
VWR® Dulbecco's Phosphate Buffered Saline (DPBS)
Catalog # VWRL0119-1000
CAS Number:
Jump to:
Discontinued
Product Details & Documents
DPBS is a balanced salt solution (BSS) used for the handling and culturing of mammalian cells.
- Used to irrigate, wash, and dilute mammalian cells
- Phosphate buffering maintains the pH in the physiological range
- Available with or without calcium and magnesium
DPBS is used to irrigate, wash, and dilute mammalian cells. Phosphate buffering maintains the pH in the physiological range. Calcium and magnesium facilitate cell binding and clumping. DPBS without these ions can be used to wash and rinse suspended cells.
Documents
Certificates
Search by Lot Number02-0119 spec DPBS 1X wo Ca wo Mg
Datasheet
Focus Industrial VWR Essential Products for Tissue Culture VWR_Life_Science_Dulbeccos_Phosphate_Buffered_Saline_DPBS_Formulation_Sheet VWR™ Focus Equipment and Instruments
Product Literature
spec_VWRL0119-0500 spec_VWRL0119-1000
Product specification
Specifications
Specifications
- Description:DPBS (1X) w/out Ca, w/out Mg
- Size:1000 ml