Chromatography Flow Control Adapters, ChemScience
Chromatography Flow Control Adapter, 50/30, Glass
Catalog # CA10822-684
Supplier: CHEM SCIENCE INC CA
Supplier #: CS-C01302
Chromatography Flow Control Adapters, ChemScience
Catalog # CA10822-684
Supplier: CHEM SCIENCE INC CA
Jump to:
Product Details & Documents
These flow control adapters with standard taper joint has a 0 to 4 mm high vacuum valve with metering tip to adjust air pressure.
Specifications
- Description:Chromatography Flow Control Adapter
- Material:Glass
- [ST] joint size:50/30