Order Entry
Canada
Orders LinkContactUsLinkComponent
Individual Slotted Killogram Weights

Individual Slotted Killogram Weights

Slotted Weight, 2 kg
Catalog # 470219-778
Supplier:   Wards
Supplier #:  PH0264B3
Individual Slotted Killogram Weights
Catalog # 470219-778
Supplier:  Wards
Individual Slotted Killogram Weights
Individual Slotted Killogram Weights

Slotted Weight, 2 kg

Price
$75.45
Log in for availability
Each
Product Details & Documents

Individual cast iron slotted weights.

  • Available individually
  • Stackable
  • Made from solid cast iron
  • Finished with a black enamel
  • Stamped with weight value

Slotted iron masses are suitable for use with sonometers or other applications, where a large load is required. The slotted masses can be hung on a hanger.

Ordering information: Hanger ordered separately.

Specifications
  • Description:
    Slotted Weight
  • Weight:
    2 kg
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Slotted Hanger Only, 0.5 kg
    Slotted Hanger Only, 0.5 kg
    470229-366
    Log in for availability
    Each - $37.90

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.