VertiKlean® MAX™ Mopping Systems, Contec®
Product 76449-910 is a functional alternative to 10816-272 but Packaging, Quantity, Manufacturer and/or specifications may vary.
Product Details & Documents
VertiKlean® MAX™ Mop was designed for applying disinfectants to both horizontal and vertical surfaces, and is ideal for general cleaning of ceilings, walls, and floors in the most critical environments. This durable, lightweight mop features a single layer hydrophilic polyurethane foam laminated to polyester microfiber fabric, backed by a semi-rigid thermoformed plate, which easily snaps onto a standard VertiKlean® mop frame.
- Hydrophilic polyurethane foam and knitted polyester mop head retains liquids
- When the mop is applied to a surface, the liquid is evenly released without dripping
- Laser cutting seals the edges of the foam and polyester, reducing the chance of fiber and particle contamination
- VertiKlean® MAX™ Mop enables the uniform application of disinfectants and cleaning solutions
- Excellent chemical and abrasion resistance
- Lasts longer than traditional sponge mops
- Compatible with a wide range of solutions
- All three sizes available with a sealed edge
- Available gamma irradiated. Validated sterile to a Sterility Assurance Level of 10⁻⁶
VertiKlean® MAX™ Mop has a slim profile for cleaning in confined spaces. Ribbed microfiber material smoothly glides over floors and textured surfaces and provides superior contamination removal. The pivoting mop frame makes it easy to clean around door and window frames as well as light fixtures, and heavy equipment. Available in three lengths to meet any cleaning needs, the smallest VertiKlean® MAX™ Mop is appropriate for cleaning tasks in confined spaces, such as cleaning and disinfecting fume hoods, isolators and mini environments.
The VertiKlean® MAX™ Mop Medium and Large are ideal for cleaning and disinfecting broad surfaces in cleanrooms and controlled environments. All three sizes are available with a sealed edge. For superior particle pick up and contamination removal, as well as maximum sorbency, choose VertiKlean® MAX™ for cleaning and disinfecting.
Specifications
- Description:QuickConnect™ Mop Frame for Regular Systems, Stainless Steel
- Size:19 cm (7.5")
Accessories
Product Information
- Catalog #LengthAvailabilityPrice
- QuickConnect™ stainless steel telescopic handleQuickConnect™ stainless steel telescopic handle33502-717127 - 234 cm (50 - 92")Log in for availabilityEach - $761.10
Product Information
- Catalog #LengthAvailabilityPrice
- QuickConnect™ anodized aluminum telescopic handleQuickConnect™ anodized aluminum telescopic handle33502-715127 - 234 cm (50 - 92")Log in for availabilityEach - $265.74
Product Information
- Catalog #LengthAvailabilityPrice
- QuickConnect™ stainless steel telescopic handleQuickConnect™ stainless steel telescopic handle33502-71641 - 76 cm (16 - 30")Log in for availabilityEach - $502.24
Product Information
- Catalog #LengthAvailabilityPrice
- QuickConnect™ anodized aluminum telescopic handleQuickConnect™ anodized aluminum telescopic handle33502-71441 - 76 cm (16 - 30")Log in for availabilityEach - $235.64