Order Entry
Canada
Orders LinkContactUsLinkComponent
Fixable Viability Kit, Zombie Violet™

Fixable Viability Kit, Zombie Violet™

Zombie Violet™ Fixable Viability Kit, 100 Tests
Catalog # CA10761-308
Supplier:   Biolegend
Supplier #:  423113
Fixable Viability Kit, Zombie Violet™
Catalog # CA10761-308
Supplier:  Biolegend
CAS Number:  
Fixable Viability Kit, Zombie Violet™
Fixable Viability Kit, Zombie Violet™

Zombie Violet™ Fixable Viability Kit, 100 Tests

CAS Number:  
Price
139,00 $
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Zombie Violet™ is an amine-reactive fluorescent dye that is non-permeant to live cells but permeant to the cells with compromised membranes

Zombie Violet™ dye is excited by the violet laser (405 nm) and has a fluorescence emission maximum of 423 nm. If using in a multi-color panel design, filter optimization may be required depending on other fluorophores used.

Thus, it can be used to assess live vs. dead status of mammalian cells. Zombie Violet™ is a polar, water soluble dye providing green fluorescence, making it suitable for multi-color detection.

Delivery information: Zombie Violet™ Fixable Viability Kit is composed of lyophilized Zombie Violet™ dye and anhydrous DMSO.

Specifications

Specifications

  • Description:
    Zombie Violet™ Fixable Viability Kit
  • Size:
    100 Tests

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.