Optical Refraction Components
Equilateral Prism, Acrylic, 75 sides x 9 mm Thick
Catalog # 470219-480
Supplier: Wards
Supplier #: PH0569A
Jump to:
Product Details & Documents
These Optical Refraction Components are made from imported optical glass or acrylic with optically worked surfaces.
- Students Measure the Refraction of Light
- Refraction Blocks, Prisms, and Lenses
- Optically Worked Surfaces
- Bubble-Free Glass
- Ideal in Conjunction with a Light Box
All angles are accurately worked with edges slightly beveled and glass free from bubbles.
These optical elements are perfect to use with a light box, so students can study and measure the refraction of light through different mediums and different shapes.
Specifications
- Description:Equilateral Prism
- Dimensions:75 sides x 9 mm Thick
- Material:Acrylic