Orders LinkContactUsLinkComponent
Optical Refraction Components
Optical Refraction Components
Rectangular Block, Glass, 75 x 50 x 12 mm
Catalog # 470219-464
Supplier:   Wards
Supplier #:  PH0566A
Optical Refraction Components
Catalog # 470219-464
Supplier:  Wards
Optical Refraction Components
Optical Refraction Components
Rectangular Block, Glass, 75 x 50 x 12 mm
Clearance
$3.25
Sale
Valid until 31/12/2026
Log in for availability
Each
Product Details & Documents
These Optical Refraction Components are made from imported optical glass or acrylic with optically worked surfaces.
  • Students Measure the Refraction of Light
  • Refraction Blocks, Prisms, and Lenses
  • Optically Worked Surfaces
  • Bubble-Free Glass
  • Ideal in Conjunction with a Light Box
All angles are accurately worked with edges slightly beveled and glass free from bubbles.
These optical elements are perfect to use with a light box, so students can study and measure the refraction of light through different mediums and different shapes.
Specifications
  • Description:
    Rectangular Block
  • Dimensions:
    75 x 50 x 12 mm
  • Material:
    Glass
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.