Orders LinkContactUsLinkComponent
VWR® Graefe Micro Hook
VWR® Graefe Micro Hook
VWR® Graefe Micro Hook, Steel, 11.4 cm (4.5"), Sharp
Catalog # 10806-340
Supplier:   VWR International
VWR® Graefe Micro Hook
Catalog # 10806-340
Supplier:  VWR International
VWR® Graefe Micro Hook
VWR® Graefe Micro Hook
VWR® Graefe Micro Hook, Steel, 11.4 cm (4.5"), Sharp
Price
$87.00
New Customer Promotional Price
Log in for availability
Each Pack of 12
Product Details & Documents
The VWR® Graefe Micro Hook is constructed from German steel.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Devices are available in sharp or blunt formats.
Specifications
  • Description:
    VWR® Graefe Micro Hook
  • Length:
    11.4 cm (4.5")
  • Material:
    Steel
  • Tip style:
    Sharp