Orders LinkContactUsLinkComponent
Innovating Science® Document Analysis: Comprehensive Lab Activities
Document Analysis: Comprehensive Lab Activities
  470219-090
 :  IS9017
Innovating Science® Document Analysis: Comprehensive Lab Activities
  470219-090
Innovating Science® Document Analysis: Comprehensive Lab Activities
Innovating Science® Document Analysis: Comprehensive Lab Activities
Document Analysis: Comprehensive Lab Activities
$368.75
Each
This kit has been designed to introduce students to the field of document analysis. Experiments in this kit cover the areas of paper analysis, hand writing analysis, detection of a forgery, recovery of mechanical/chemical erasures, hidden message discovery and ink analysis. Students will make use of techniques like microscopy, chromatography and fuming to complete the activities. Kit contains 10 unique sets of materials for 10 groups. Teacher’s Manual and Student Study Guide copymasters are included.
  • Kit contains 10 unique sets of materials for 10 groups
  • Teacher’s Manual and Student Study Guide copymasters are included
Experiments in this kit cover the areas of paper analysis, hand writing analysis, detection of a forgery, recovery of mechanical/chemical erasures, hidden message discovery and ink analysis. Students will make use of techniques like microscopy, chromatography and fuming to complete the activities.
  • Description:
    Document Analysis: Comprehensive Lab Activities
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.