pH Buffer Solutions, NIST
Bottle, 500 ml, 1.68
Catalog # 10804-868
Supplier: Mettler Toledo
Supplier #: 30258811
Jump to:
Product Details & Documents
These buffer solutions are used for calibration and verification of measurement system.
- Shelf life: 24 months
- Uncertainty: ±0.02 pH
Certifications: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Specifications
Specifications
- Pack type:Bottle
- Size:500 ml
- pH:1.68
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



