Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Solutions, NIST

pH Buffer Solutions, NIST

Bottle, 500 ml, 5.00
Catalog # 10804-850
Supplier:   Mettler Toledo
Supplier #:  30258823
pH Buffer Solutions, NIST
Catalog # 10804-850
Supplier:  Mettler Toledo
CAS Number:  
pH Buffer Solutions, NIST
pH Buffer Solutions, NIST

Bottle, 500 ml, 5.00

CAS Number:  
Price
$49.57
New Customer Promotional Price
Log in for availability
Each (500ml)
Product Details & Documents

These buffer solutions are used for calibration and verification of measurement system.

  • Shelf life: 24 months
  • Uncertainty: ±0.02 pH

Certifications: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 ml
  • pH:
    5.00

Specifications