Screw Caps, MicroSolv™
PP, Silicone Rubber / PTFE Pre-Slit, 13 - 425 mm, White, Screw cap
Catalog # 10804-132
Supplier: MicroSolv Technology Corp
Supplier #: 9504S-30C-W
Jump to:
Product Details & Documents
These screw caps are designed for use with 13-425 cap size, 15x45mm vials.
Specifications
- Cap Size:13 - 425 mm
- Cap type:Screw cap
- Septum:Silicone Rubber / PTFE Pre-Slit
- Material:PP
- Cap color:White