Screw Caps, MicroSolv™
PP, Red Orange Natural Rubber/ Transparent TFE, 13 - 425 mm, Black, Screw cap
Catalog # 10804-128
Supplier: MicroSolv Technology Corp
Supplier #: 9504S-80C-BK
Jump to:
Product Details & Documents
These screw caps are designed for use with 13-425 cap size, 15x45mm vials.
Specifications
- Cap Size:13 - 425 mm
- Cap type:Screw cap
- Septum:Red Orange Natural Rubber/ Transparent TFE
- Material:PP
- Cap color:Black