Order Entry
Canada
Orders LinkContactUsLinkComponent
Scotch Conductive Tapes, Electron Microscopy Sciences

Scotch Conductive Tapes, Electron Microscopy Sciences

Aluminum Tape, 1"
Catalog # 102091-341
Supplier #:  77798
Scotch Conductive Tapes, Electron Microscopy Sciences
Catalog # 102091-341
Scotch Conductive Tapes, Electron Microscopy Sciences
Scotch Conductive Tapes, Electron Microscopy Sciences

Aluminum Tape, 1"

Price
$290.94
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Copper and aluminum conductive tapes with continuous long term usage up to 155 °C

  • Functional days-weeks usage up to 175 °C
  • Copper double sided available

We offer two types of foil: Aluminum foil tape (Scotch 1170) and copper foil tape (Scotch 1181) with conductive adhesive. 3” core, 1/4" (6.35 mm) width×54 ft (16.5 m) long.

Documents

Specifications
  • Width:
    1"
  • Description:
    Aluminum Tape

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.