Orders LinkContactUsLinkComponent
Dropping Bottles
Dropping Bottles
Clear, 30ml
Catalog # 470217-646
Supplier:   Wards
Supplier #:  CH0187A
Dropping Bottles
Catalog # 470217-646
Supplier:  Wards
Dropping Bottles
Dropping Bottles
Clear, 30ml
Clearance
$3.00
Sale
Valid until 31/12/2026
Log in for availability
Each
Product Details & Documents
Dropping bottles are available in clear or amber formats.
  • Screw cap included
  • Dropper included
Bottles are made of soda glass.
Specifications
  • Description:
    Clear, 30ml