$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
CAPS SCREW SOFT GUARD SIL/PTFE RD CS
10803-664
:
MicroSolv Technology Corp
CAPS SCREW SOFT GUARD SIL/PTFE RD CS
10803-664
:
MicroSolv Technology Corp
:
9502S-E-10C-RM
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning