Caps, BASIK™, MicroSolv
PE, Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit, 11 mm, Green, Snap cap
Catalog # 10803-606
Supplier: MicroSolv Technology Corp
Supplier #: 9502C-E-30G-M
Jump to:
Product Details & Documents
Specifications
- Cap Size:11 mm
- Cap type:Snap cap
- Septum:Soft-Guard, White Silicone Rubber / Red PTFE Pre-Slit
- Material:PE
- Cap color:Green