Order Entry
Canada
Orders LinkContactUsLinkComponent
Triangular Base and Rod Assembly

Triangular Base and Rod Assembly

170 mm, 50 cm
Catalog # 470165-338
Supplier:   Wards
Supplier #:  TRISTD1
Triangular Base and Rod Assembly
Catalog # 470165-338
Supplier:  Wards
Triangular Base and Rod Assembly
Triangular Base and Rod Assembly

170 mm, 50 cm

Price
$50.40
Log in for availability
Each
Product Details & Documents

Sturdy assembly for all of your laboratory needs.

  • Heavy-duty cast iron construction
  • Two sizes available

Triangular-shaped, cast iron base provides stability. Base is 23mm wide (radius: 13cm). Each size includes a threaded steal rod.

Specifications
  • Rod length:
    50 cm
  • Base dimensions:
    170 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.