Order Entry
Canada
Orders LinkContactUsLinkComponent
Corning® Plain Micro Slides, Corning

Corning® Plain Micro Slides, Corning

Plain slides, 0.9 - 1.0 mm, 75×50 mm (3×2")
Catalog # 89091-009
Supplier:   Corning
Supplier #:  2947-75X50
Corning® Plain Micro Slides, Corning
Catalog # 89091-009
Supplier:  Corning
Corning® Plain Micro Slides, Corning
Corning® Plain Micro Slides, Corning

Plain slides, 0.9 - 1.0 mm, 75×50 mm (3×2")

Price
$779.00
New Customer Promotional Price
Log in for availability
Case of 720
Product Details & Documents

For use in nonsterile cytology and pathology applications

Made of water-white glass for maximum clarity. Precleaned and ready for use. Inspected for chips and rough edges before packaging.

Ordering information: Packaged 1/2 gross per box. Slides are packed vertically for easy lift out.

Documents

Specifications
  • Description:
    Plain slides
  • L×W:
    75×50 mm (3×2")
  • Thickness:
    0.9 - 1.0 mm