Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
BAG SPECM 12X15 CLR BIO 3 WL ZIP ABSORB
10789-516
:
Elkay Plastics
BAG SPECM 12X15 CLR BIO 3 WL ZIP ABSORB
10789-516
:
Elkay Plastics
:
LABZ1215BA
Elkay Seal-N-Rip Specimen and Large Transport Bags Flyer
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Universal Pipette Tips
: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH