Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BAG SPECM 8X10 CLR BIO 3 WALL ZIP ABSORB
Catalog # 10789-512
Supplier:
Elkay Plastics
Expand image
BAG SPECM 8X10 CLR BIO 3 WALL ZIP ABSORB
Catalog # 10789-512
Supplier:
Elkay Plastics
Supplier Number:
LABZ810BA
Related Information
Documentation
Product Literature
Elkay Seal-N-Rip Specimen and Large Transport Bags Flyer
Documentation
Frequently Bought Together
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International