Axygen® Automation Tips for Molecular Devices FLIPR Tetra, Corning
384-Well Format, 30 µl, Black, Racked
Supplier: Corning
Supplier #: FT-384-BK-R
Axygen® Automation Tips for Molecular Devices FLIPR Tetra, Corning
Catalog # CA10770-592
Supplier: Corning
Jump to:
Product Details & Documents
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Designed for use in molecular devices FLIPR tetra automation systems.
- Manufactured to stringent specifications under strict process controls
- Assembled via automation to ensure consistent performance and quality
- Certified RNase-/DNase-free and non pyrogenic
- Black and clear tips in 384-well format
Documents
Certificates
Search by Lot NumberCorning Automation Tip Compatibility Chart Corning Axygen Automation Tips
Product Literature
Specifications
- Volume:30 µl
- Tip color:Black
- Packed:Racked
- Tip style:384-Well Format