Order Entry
Canada
Orders LinkContactUsLinkComponent
SpillStopper Work Surfaces, Labconco | Avantor

SpillStopper Work Surfaces, Labconco

Work Surface-LS,RS, 36", 48", Left side and right side
  10769-394
 :   Labconco
Fournisseur #:  9501422
SpillStopper Work Surfaces, Labconco
  10769-394
 :  Labconco
SpillStopper Work Surfaces, Labconco
SpillStopper Work Surfaces, Labconco

Work Surface-LS,RS, 36", 48", Left side and right side

 
$3,990.00
   
Each
La quantité minimale de commande est de 1
 

SpillStopper Work Surfaces are molded from a special formulation of corrosion-resistant epoxy resins

Dished and contoured to conform to the interior liner of Protector Hoods. Front edge has a large radius to aerodynamically direct airflow into the hood. Pre-drilled 1.5" (5.8 cm) diameter holes for venting and 2.5" (6.4 cm) diameter holes for plumbing pass-through.

Ordering information: May be ordered with a pre-cut 15.2 x 7.6 cm (6 x 3") oval cupsink cutout. Cupsink is sold separately. SpillStopper Work Surfaces provide support for Protector Premier, XStream, XL, Echo and Airo Laboratory Hoods.

 
  • Description:
    Work Surface-LS,RS
  • Depth:
    36"
  • Width:
    48"
  • CuPSInk cutout:
    Left side and right side
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.