Orders LinkContactUsLinkComponent
Nitrification Control Test Kit, Hach
Nitrification Control Test Kit, Hach
Free ammonia, 0.02 - 0.50 mg/L NH₃-N, monochLoramine, 0.04 - 4.50 mg/L CL₂, 0 - 0.350 mg/L NO₂-N, 50 - 100
Catalog # CA89500-778
Supplier:   Hach
Supplier #:  2882200
Nitrification Control Test Kit, Hach
Catalog # CA89500-778
Supplier:  Hach
CAS Number:  
Nitrification Control Test Kit, Hach
Nitrification Control Test Kit, Hach
Free ammonia, 0.02 - 0.50 mg/L NH₃-N, monochLoramine, 0.04 - 4.50 mg/L CL₂, 0 - 0.350 mg/L NO₂-N, 50 - 100
CAS Number:  
Price
7 549,33 $
Log in for availability
Each
Product Details & Documents
The Nitrification Control Kit is used for the routine monitoring of water quality parameters that contribute to or indicate nitrification is occurring in chloraminated water systems. This portable and versatile kit includes a colorimeter and reagents for Free Ammonia and Monochloramine, Nitrite, and Total Chlorine tests.
Used for the routine monitoring of water quality parameters that contribute to or indicate nitrification is occurring in chloraminated water systems
Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Versatile kit contains the colorimeter and reagents to perform tests for Free Ammonia, Monochloramine, Nitrite, Total Chlorine (low range and high range).
Documents
Specifications
Specifications
  • Range:
    Free ammonia, 0.02 - 0.50 mg/L NH₃-N, monochLoramine, 0.04 - 4.50 mg/L CL₂, 0 - 0.350 mg/L NO₂-N
  • No. of tests:
    50 - 100
  • Cat. No.:
    CA89500-778
  • Platform:
    PP, S