Orders LinkContactUsLinkComponent
Model ASC-1 Ascorbic Acid Test Kit, Hach
Model ASC-1 Ascorbic Acid Test Kit, Hach
Iodometric, 10 - 200 mg/L ascorbic acid, 100
Catalog # CA89500-718
Supplier:   Hach
Supplier #:  2308100
Model ASC-1 Ascorbic Acid Test Kit, Hach
Catalog # CA89500-718
Supplier:  Hach
CAS Number:  
Model ASC-1 Ascorbic Acid Test Kit, Hach
Model ASC-1 Ascorbic Acid Test Kit, Hach
Iodometric, 10 - 200 mg/L ascorbic acid, 100
CAS Number:  
Price
$352.31
Log in for availability
Each
Product Details & Documents
Model ASC-1 Ascorbic Acid 100-Test Kit was developed in cooperation with beverage producers and uses the drop-count titration method to measure ascorbic acid (vitamin C)
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Utilizes drop count titration with a range of 10-200 mg/L. This kit utilizes drop-count titration simply and economically. It contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
Ordering information: Sample measuring tube, flask, reagents, instructions and carrying case included.
Documents
Specifications
Specifications
  • Chemical method:
    Iodometric
  • Range:
    10 - 200 mg/L ascorbic acid
  • No. of tests:
    100
  • Cat. No.:
    CA89500-718
  • Platform:
    DCT
  • Method:
    Drop Count Titration/ Iodometric