Order Entry
Canada
Orders LinkContactUsLinkComponent
 

pH Buffer Solutions, Hach

±0.02, 25 °C, 15 mL, 7.00, Yellow
Catalog # CA10199-506
Supplier:   Hach
 :  2283536
pH Buffer Solutions, Hach
Catalog # CA10199-506
Supplier:  Hach
CAS Number:  
pH Buffer Solutions, Hach
pH Buffer Solutions, Hach

±0.02, 25 °C, 15 mL, 7.00, Yellow

CAS Number:  
Price
$22.46
Log in for availability
Each
Product Details & Documents

For pH meter standardization

Available colorless, or color-coded for easy identification.

Specifications

Specifications

  • Color:
    Yellow
  • Size:
    15 mL
  • Reference temperature:
    25 °C
  • pH:
    7.00
  • pH accuracy:
    ±0.02

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.