Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Solutions, Hach

pH Buffer Solutions, Hach

±0.02, 25 °C, 50 mL, 4.01, Red
Catalog # CA10199-500
Supplier:   Hach
Supplier #:  2283426
pH Buffer Solutions, Hach
Catalog # CA10199-500
Supplier:  Hach
CAS Number:  
pH Buffer Solutions, Hach
pH Buffer Solutions, Hach

±0.02, 25 °C, 50 mL, 4.01, Red

CAS Number:  
Price
$34.77
Log in for availability
Each
Product Details & Documents

For pH meter standardization

Available colorless, or color-coded for easy identification.

Specifications

Specifications

  • Color:
    Red
  • Size:
    50 mL
  • Reference temperature:
    25 °C
  • pH:
    4.01
  • pH accuracy:
    ±0.02

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.