Orders LinkContactUsLinkComponent
ASTM Class 7 Hooked Weights
ASTM Class 7 Hooked Weights
0.5 N
Catalog # 10754-374
Supplier:   Troemner
Supplier #:  30390782
ASTM Class 7 Hooked Weights
Catalog # 10754-374
Supplier:  Troemner
ASTM Class 7 Hooked Weights
ASTM Class 7 Hooked Weights
0.5 N
Price
$59.53
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
ASTM hooked weights for weighing operations in physical and chemical classrooms, such as force measuring.
  • 303 stainless steel
  • ASTM Class 7
All surfaces are satin finish and meet or exceed ANSI/ASTM E617 tolerance and design restrictions.
Specifications
  • Weight:
    0.5 N