Order Entry
Canada
ContactUsLinkComponent
Specimen Container
HistoTainer™ Prefilled Specimen Containers, with 10% Neutral Buffered Formalin, Simport Scientific
Catalog # CA10662-322
Supplier:  Simport Scientific
undefined
HistoTainer™ Prefilled Specimen Containers, with 10% Neutral Buffered Formalin, Simport Scientific
Catalog # CA10662-322
Supplier:  Simport Scientific
Supplier Number:  M961-40FW

Specifications

  • Capacity:
    40 ml
  • Description:
    HistoTainer™ II Specimen Containers
  • Cat. no.:
    CA10662-322
  • Supplier No.:
    M961-40FW

Specifications

About this item

Shatter resistant and leak proof polypropylene containers are designed for collection, transport, and storage of histology specimens

The HistoTainer™ I includes a tamper evident leak-proof screw cap. A special push-tab mechanism is integrated in the container. These containers may also be used without using the tamper evident locking mechanism. The HistoTainer™ II includes a regular leak-proof screw cap.

Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used in manufacture are free from latex. Containers are 95kPa compliant. Available in multiple sizes from 20 to 120mL. Containers are made of polypropylene. Closures are made of polyethylene.

Container lids are ribbed for easy opening and the jars are stackable for convenient storage. The HistoTainer™ Prefilled Specimen Containers are available in two formats.

Certifications: CE marked.
Ordering information: For color coding purposes, use a Capinsert™ (see T345 Series) on top of closure. Ten different colors are available. HistoTainer™ I containers are available with magenta cap. HistoTainer™ II containers are available with white cap.
Frequently Bought Together