VWR® Flat-Bottom Boiling Flasks
300 ml, 24/40
Supplier: VWR International
Jump to:
Product Details & Documents
The flasks come with uniform wall thickness which makes these flasks ideal for heating.
- Borosilicate glass fabrication
- Good chemical resistance
Durable product has good temperature and chemical resistance.
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. As per ASTM specification E 1403, Type 1 Class 4.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:300 ml
- Neck size:24/40