$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
GSBP-FFAP 10M 0.10MM 0.10UM
Catalog # 10499-520
Supplier:
General Separation Technologies, Inc.
Expand image
GSBP-FFAP 10M 0.10MM 0.10UM
Catalog # 10499-520
Supplier:
General Separation Technologies, Inc.
Supplier Number:
2110-1010
Frequently Bought Together
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International