Certipur® Buffer Solution Sachet Sets, MilliporeSigma
Certipur® buffer solution, set 2, Sachet, 30 x 30 mL, 4.01/7.00/10.00
: 1.99006.0001
Certipur® Buffer Solution Sachet Sets, MilliporeSigma
Catalog # CA1.99006.0001
CAS Number:
Jump to:
Discontinued
Product Details & Documents
30 ml buffers sachets - ideal for the calibration of pH meters.
- Practical 30 mL sachets – one use per sachet – no risk of contamination
- Solutions are always fresh and ready to use
- Batch traceable to NIST
- Reference temperature: 25°C
- Precision: ±0.015 pH units max.
Specifications
Specifications
- Pack type:Sachet
- Description:Certipur® buffer solution, set 2
- Size:30 x 30 mL
- pH:4.01/7.00/10.00
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



