Flow Indicators, Bürkle
Product Details & Documents
These paddle wheel flow indicators give a clear visualisation of the flow rates of liquids or gas in tubing
LiquiMobil flow indicator made of SAN is mainly suitable for cooling systems and helps to reduce water consumption considerably because the cooling water fl ow can be adjusted better. Not suitable for aggressive substances. Maximum operating temperature LiquiMobil SAN is +30 °C.
CheMobil PMP shows reasonable chemical resistance, which makes it possible to use with a great number of fluids. CheMobil is not however appropriate for strong acids, bases and solvents. CheMobil is steam sterilisable. Maximum operating temperature +60 °C.
The flow indicator can operate in either direction of flow and can be mounted at any angle. The paddle wheel impeller starts to rotate at about 150 ml/min. Colour coding - red, black or blue
Documents
Specifications
- Type:CheMobil
- Color:Black
- Material:PMP
- Tube diameter:6 - 11 mm