Ward's® Plastic Slide Boxes
25-Capacity Slide Box
Catalog # 470151-632
Supplier: Ward's Science
Supplier #: 481010-R
Jump to:
Product Details & Documents
The plastic slide box protects 1x3" slides and won't warp when temperature and humidity vary.
- Plastic construction
- Foam-lined bottom
- Numbered insert for easy organization of slides
The 25-capacity and 100-capacity boxes have fully removable lid latches with a plastic clip. The 10-capacity boxes are hinged, with a snap-close mechanism.
Specifications
- Description:25-Capacity Slide Box