Orders LinkContactUsLinkComponent
Ward's® Plastic Slide Boxes
Ward's® Plastic Slide Boxes
100-Capacity Slide Box
Catalog # 470151-630
Supplier:   Ward's Science
Supplier #:  481030-R
Ward's® Plastic Slide Boxes
Catalog # 470151-630
Supplier:  Ward's Science
Ward's® Plastic Slide Boxes
Ward's® Plastic Slide Boxes
100-Capacity Slide Box
Price
$24.65
Log in for availability
Each Case of 6
Product Details & Documents
The plastic slide box protects 1x3" slides and won't warp when temperature and humidity vary.
  • Plastic construction
  • Foam-lined bottom
  • Numbered insert for easy organization of slides
The 25-capacity and 100-capacity boxes have fully removable lid latches with a plastic clip. The 10-capacity boxes are hinged, with a snap-close mechanism.
Specifications
  • Description:
    100-Capacity Slide Box