Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Schlenk Tubes, Modified, ChemScience

200 ml, Flask Modified Schlenk Tube, 19/22, PTFE
Catalog # CA10204-454
Supplier:   CHEM SCIENCE INC CA
 :  CS-F0909200
Schlenk Tubes, Modified, ChemScience
Catalog # CA10204-454
Schlenk Tubes, Modified, ChemScience
Schlenk Tubes, Modified, ChemScience

200 ml, Flask Modified Schlenk Tube, 19/22, PTFE

Price
$204.34
Log in for availability
Case of 1
Product Details & Documents

Choice of either 2 mm glass stopcock, 2 mm Teflon® stopcock or 0 to 4 mm high vacuum valve.

Specifications
  • Capacity:
    200 ml
  • Description:
    Flask Modified Schlenk Tube
  • [ST] joint size:
    19/22
  • Stopcock type:
    PTFE