Orders LinkContactUsLinkComponent
Stopcocks, PTFE Straight Bore, Chemscience
Stopcocks, PTFE Straight Bore, Chemscience
Stopcock, PTFE straight bore, 2 mm, 11/25
Catalog # CA10202-960
Supplier:   CHEM SCIENCE INC CA
Supplier #:  CS-S0101502
Stopcocks, PTFE Straight Bore, Chemscience
Catalog # CA10202-960
Stopcocks, PTFE Straight Bore, Chemscience
Stopcocks, PTFE Straight Bore, Chemscience
Stopcock, PTFE straight bore, 2 mm, 11/25
Price
$84.22
Log in for availability
Case of 1
Product Details & Documents
With 1:5 PTFE plug and a threaded pressure nut.
Stopcock barrels are polished to prevent PTFE plug from scratch.
Specifications
  • Description:
    Stopcock, PTFE straight bore
  • Bore size:
    2 mm
  • Plug size:
    11/25