Filtering Flasks, Heavy Wall
Filtering Flask, Heavy Wall, 500 ml, 24/40
Catalog # CA10202-246
Supplier: CHEM SCIENCE INC CA
: CS-F0155500
Jump to:
Product Details & Documents
Erlenmeyer shaped filtering flask with standard taper outer joint.
- Heavy wall design
Specifications
- Capacity:500 ml
- Description:Filtering Flask, Heavy Wall
- Joint size:24/40