Filtering Flasks, Heavy Wall
Filtering Flask, Heavy Wall, 250 ml, 19/22
Catalog # CA10202-242
Supplier: CHEM SCIENCE INC CA
Supplier #: CS-F0155251
Jump to:
Product Details & Documents
Erlenmeyer shaped filtering flask with standard taper outer joint.
- Heavy wall design
Specifications
- Capacity:250 ml
- Description:Filtering Flask, Heavy Wall
- Joint size:19/22