Orders LinkContactUsLinkComponent
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
25 ml, Round Bottom Flask, Threaded Inlet, 14/20
Catalog # CA10201-810
Supplier:   CHEM SCIENCE INC CA
Supplier #:  CS-F0141425
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Catalog # CA10201-810
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, ChemScience
25 ml, Round Bottom Flask, Threaded Inlet, 14/20
Price
$99.77
Log in for availability
Case of 1
Product Details & Documents
All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
  • An outer diameter between 5 and 7.5 mm
  • Heavy wall design
With a threaded side port for insertion of thermometer or bleed tubes.
Specifications
  • Capacity:
    25 ml
  • Description:
    Round Bottom Flask, Threaded Inlet
  • Joint size:
    14/20