Orders LinkContactUsLinkComponent
Solution Basin, Sklar
Solution Basin, Sklar
Stainless Steel Solution Basin, 7 qt., 13 5/8" x 4 5/8"
Catalog # 10192-810
Supplier:   Sklar
Supplier #:  10-1505
Solution Basin, Sklar
Catalog # 10192-810
Supplier:  Sklar
Solution Basin, Sklar
Solution Basin, Sklar
Stainless Steel Solution Basin, 7 qt., 13 5/8" x 4 5/8"
Price
$323.46
New Customer Promotional Price
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Solution Basins are versatile products that may be used for mixing and containing solutions, capturing fluids, or a receptacle for soiled sponges or dressings.
They are available in multiple sizes so as to accomodate a variety of procedures in a variety of settings.
Specifications
  • Volume:
    7 qt.
  • Description:
    Stainless Steel Solution Basin
  • Dimensions:
    13 5/8" x 4 5/8"
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)