Orders LinkContactUsLinkComponent
Solution Basins, Sklar
Solution Basins, Sklar
Solution Bassin, 7 qt., Plastic, 13 5/8 x 4 5/8", Blue
Catalog # 10192-808
Supplier:   Sklar
Supplier #:  10-1504-12
Solution Basins, Sklar
Catalog # 10192-808
Supplier:  Sklar
Solution Basins, Sklar
Solution Bassin, 7 qt., Plastic, 13 5/8 x 4 5/8", Blue
Price
$793.53
New Customer Promotional Price
Log in for availability
Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Autoclavable and reusable solution basins made of plastic. Can be used for mixing and containing solutions, capturing fluids, or a receptacle for soiled sponges or dressings.
Specifications
  • Capacity:
    7 qt.
  • Color:
    Blue
  • Description:
    Solution Bassin
  • Dimensions:
    13 5/8 x 4 5/8"
  • Material:
    Plastic
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)