pH Buffer Solutions, Hach
±0.02, 25 °C, 50 mL, 10.01, Blue
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
For pH meter standardization
Available colorless, or color-coded for easy identification.
Specifications
Specifications
- Color:Blue
- Size:50 mL
- Reference temperature:25 °C
- pH:10.01
- pH accuracy:±0.02



