Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Solutions, Hach

pH Buffer Solutions, Hach

±0.02, 25 °C, 15 mL, 4.01, Red
Catalog # 10199-502
pH Buffer Solutions, Hach
Catalog # 10199-502
Supplier:  
CAS Number:  
pH Buffer Solutions, Hach
pH Buffer Solutions, Hach

±0.02, 25 °C, 15 mL, 4.01, Red

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BUFFER SOLN RED PH4.01 15ML Catalog #CA10199-502

Product CA10199-502 is a replacement for 10199-502 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

For pH meter standardization

Available colorless, or color-coded for easy identification.

Specifications

Specifications

  • Color:
    Red
  • Size:
    15 mL
  • Reference temperature:
    25 °C
  • pH:
    4.01
  • pH accuracy:
    ±0.02

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.